Assay Details
| Gene Symbol: Gm5416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082542.1 | NP_001076011.1 | 2 - 3 | 173 | 136 | |
| GenBank mRNA | BC115936.1 | - | 2 - 3 | 172 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997421 | NM_001082542.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997421 | BC115936.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12344 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329843 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8969 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997421 | NM_001082542.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997421 | BC115936.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12651540 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12651541 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12651531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12735506 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12735512 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12735507 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12651536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30674261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12912493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12735506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12764574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12735507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12735513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12960555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30674240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12936230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12936234 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10435507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435506 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10435504 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600004850.mm.1 | NM_001082542.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004850.mm.1 | NM_001082542.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004850.mm.1 | TC1600000510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600004851.mm.1 | NM_001082542.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004851.mm.1 | NM_001082542.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004851.mm.1 | TC1600000510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600004852.mm.1 | NM_001082542.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004852.mm.1 | NM_001082542.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600004852.mm.1 | TC1600000510.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2794741 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2794741 | - | - |

Gm5416
Genomic Map