Assay Details
| Gene Symbol: Rbpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080927.2 | NP_001074396.1 | 4 - 5 | 535 | 130 | |
| NM_001080928.1 | NP_001074397.1 | 4 - 5 | 421 | 130 | ||
| NM_001277116.1 | NP_001264045.1 | 6 - 7 | 617 | 130 | ||
| NM_009035.5 | NP_033061.3 | 4 - 5 | 437 | 130 | ||
| GenBank mRNA | AK028363.1 | - | 6 - 7 | 595 | 130 | |
| AK080359.1 | - | 4 - 5 | 330 | 130 | ||
| AK090006.1 | - | 4 - 5 | 323 | 130 | ||
| AK132145.1 | - | 4 - 5 | 379 | 130 | ||
| AK139087.1 | - | 4 - 5 | 322 | 130 | ||
| AK145187.1 | - | 4 - 5 | 329 | 130 | ||
| AK152746.1 | - | 4 - 5 | 437 | 130 | ||
| AK153163.1 | - | 5 - 6 | 325 | 130 | ||
| BC035299.1 | - | 2 - 3 | 223 | 130 | ||
| BC051387.1 | - | 4 - 5 | 366 | 4460290 | 130 | |
| CD540070.1 | - | 4 - 5 | 375 | 130 | ||
| S63463.1 | - | 4 - 5 | 438 | 130 | ||
| X17459.1 | - | 4 - 5 | 433 | 130 | ||
| Y18248.1 | - | 1 - 2 | 50 | 130 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbpj
Genomic Map