Assay Details
| Gene Symbol: Kcnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008430.2 | NP_032456.2 | 2 - 3 | 1108 | 100 | |
| GenBank mRNA | AF033017.1 | - | 2 - 3 | 1040 | 100 | |
| AK137224.1 | - | 2 - 3 | 1022 | 100 | ||
| BC003729.1 | - | 2 - 3 | 805 | 3592756 | 100 | |
| BG088333.2 | - | 1 - 2 | 398 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnk1
Genomic Map