| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100019715.mm.1 |
NM_001199060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019715.mm.1 |
NM_001199060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019715.mm.1 |
NM_001199061.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019715.mm.1 |
NM_001199061.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019715.mm.1 |
NM_021312.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019715.mm.1 |
NM_021312.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019715.mm.1 |
TC0100002411.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100019716.mm.1 |
NM_001199060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019716.mm.1 |
NM_001199060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019716.mm.1 |
NM_001199061.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019716.mm.1 |
NM_001199061.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019716.mm.1 |
NM_021312.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019716.mm.1 |
NM_021312.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100019716.mm.1 |
TC0100002411.mm.1 |
MTA TC |