Assay Details
| Gene Symbol: Olfr1388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146467.1 | NP_666678.1 | 1 | 153 | 78 | |
| GenBank mRNA | BC104274.1 | - | 1 | 152 | 78 | |
| BC104275.1 | - | 1 | 153 | 40041223 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2088940 | BC104274.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2088940 | BC104275.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2088940 | NM_146467.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2088941 | BC104274.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2088941 | BC104275.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2088941 | NM_146467.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131644 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2088940 | BC104274.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2088940 | BC104275.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2088940 | NM_146467.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2088941 | BC104274.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2088941 | BC104275.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2088941 | NM_146467.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P221833 | BC104275.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P221833 | NM_146467.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27782990 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131644 | - | - |
| Whole Mouse Genome G4122A | A_52_P221833 | BC104275.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P221833 | NM_146467.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137050 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131644 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09687181 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09687199 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09687183 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09506027 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09407643 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09407644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27782931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09687197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09687195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09690918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09397892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09690900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09687184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09687194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09690909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09637980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27782972 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375558 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10375557 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 423363 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6040717 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6040717 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040717 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2655044 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2655044 | - | - |
| mouse-6 v1.1 (Illumina) | scl41628.1.1_72-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040717 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41628.1.1_72-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040717 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2655044 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2655044 | - | - |

Olfr1388
Genomic Map