Assay Details
| Gene Symbol: Gpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290742.1 | NP_001277671.1 | 2 - 3 | 584 | 106 | |
| NM_133884.2 | NP_598645.2 | 2 - 3 | 653 | 106 | ||
| GenBank mRNA | BC018407.1 | - | 2 - 3 | 646 | 4015340 | 106 |
| CN721622.1 | - | 2 - 3 | 255 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpn2
Genomic Map