Assay Details
| Gene Symbol: Olfr1122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147031.1 | NP_667242.1 | 1 | 713 | 98 | |
| GenBank mRNA | BC119331.1 | - | 1 | 764 | 8734080 | 98 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377492 | NM_147031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377492 | BC119331.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377492 | NM_147031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377492 | BC119331.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443758 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113744 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377492 | NM_147031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377492 | BC119331.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377492 | NM_147031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377492 | BC119331.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P377492 | NM_147031.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P377492 | BC119331.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P377492 | NM_147031.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P377492 | BC119331.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405779 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113744 | - | - |
| Whole Mouse Genome G4122A | A_52_P377492 | NM_147031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377492 | BC119331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377492 | NM_147031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377492 | BC119331.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125835 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113744 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01588828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01588824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01588837 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01588830 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01588842 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01588825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01939624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01939609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01868801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01868799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01588830 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10473536 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473534 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473533 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200010037.mm.1 | TC0200001289.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 337409 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2370161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002370161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241550 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241550 | - | - |
| mouse-6 v1.1 (Illumina) | scl4915.1.1_45-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2370161 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4915.1.1_45-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370161 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1241550 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241550 | - | - |

Olfr1122
Genomic Map