Assay Details
| Gene Symbol: Olfr960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146279.1 | NP_666391.1 | 1 | 267 | 133 | |
| GenBank mRNA | BC132468.1 | - | 1 | 287 | 40130745 | 133 |
| BC132470.1 | - | 1 | 287 | 40130747 | 133 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr960
Genomic Map