Assay Details
| Gene Symbol: Nkx2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010919.2 | NP_035049.1 | 2 | 2001 | 61 | |
| GenBank mRNA | AK141002.1 | - | 2 | 2006 | 61 | |
| U31566.1 | - | 2 | 1978 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nkx2-2
Genomic Map