Assay Details
| Gene Symbol: Eras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181548.2 | NP_853526.1 | 2 | 1028 | 76 | |
| GenBank mRNA | AB093573.1 | - | 2 | 1028 | 76 | |
| AK141212.1 | - | 2 | 922 | 76 | ||
| Gene Symbol: Pcsk1n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013892.3 | NP_038920.2 | 3 | 76 | ||
| GenBank mRNA | BE986911.1 | - | 1 | 101 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
small GTPase mediated signal transduction
biological_process
response to dietary excess
neuropeptide signaling pathway
response to cold
negative regulation of endopeptidase activity
peptide hormone processing
nucleotide binding
GTP binding
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
signal transduction
small GTPase mediated signal transduction
biological_process
response to dietary excess
neuropeptide signaling pathway
response to cold
negative regulation of endopeptidase activity
peptide hormone processing
nucleotide binding
GTP binding
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | NM_181548.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | AB093573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | AK141212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | BC119277.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | BC119281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | NM_181548.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | AB093573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | AK141212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | BC119277.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275649 | BC119281.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99867 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165770 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | NM_181548.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | AB093573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | AK141212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | BC119277.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | BC119281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | NM_181548.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | AB093573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | AK141212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | BC119277.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275649 | BC119281.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | NM_181548.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | AB093573.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | AK141212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | BC119277.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | BC119281.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | NM_181548.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | AB093573.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | AK141212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | BC119277.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275649 | BC119281.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32240942 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165770 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09227167 | - | - |
| Whole Mouse Genome G4122A | A_52_P275649 | NM_181548.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275649 | AB093573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275649 | AK141212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275649 | BC119277.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275649 | BC119281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275649 | NM_181548.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275649 | AB093573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275649 | AK141212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275649 | BC119277.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275649 | BC119281.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122893 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165770 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14557010 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14556996 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14556995 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14830408 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14556989 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14556987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32241017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32240947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15026693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15027396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14556991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14557009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14556996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15026692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14556990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32240942 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10603383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10603386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10603385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10603384 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456511_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10603382 | - | - |
| Mouse Genome 430 2.0 Array | 1456511_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1456511_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 353283_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 353283_PM_at | - | - |
| Mouse Expression Set 430 | 1456511_PM_x_at | - | - |
| Mouse Expression Set 430 | 1456511_x_at | - | - |
| Murine Genome U74v2 Set | 165826_PM_i_at | - | - |
| Murine Genome U74v2 Set | 165826_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456511_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456511_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456511_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456511_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 302319 | - | - |
| ABI Mouse Genome Survey Microarray | 868267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3610577 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610577 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2725563 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2725563 | - | - |
| mouse-6 v1.1 (Illumina) | scl54441.2.1_273-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610577 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54441.2.1_273-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610577 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2844838 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2844838 | - | - |

Eras
Genomic Map