Assay Details
| Gene Symbol: Yy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098723.1 | NP_001092193.1 | 1 | 1061 | 135 | |
| GenBank mRNA | AK161451.1 | - | 1 | 1063 | 135 | |
| Gene Symbol: Mbtps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036071.1 | - | 6 | 1913 | 135 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184197 | NM_001098723.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184197 | AK161451.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184197 | AK036071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184197 | NM_001098723.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184197 | AK161451.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184197 | AK036071.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32767059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14943487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15219546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14943456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14627199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15219545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15184106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14683232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32767066 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10607549 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607548 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607547 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10607539 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00020368.mm.1 | NM_001098723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020368.mm.1 | NM_001098723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020368.mm.1 | TC0X00003250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00020371.mm.1 | NM_001098723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020371.mm.1 | NM_001098723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020371.mm.1 | TC0X00003250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00020370.mm.1 | NM_001098723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020370.mm.1 | NM_001098723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020370.mm.1 | TC0X00003250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00020369.mm.1 | NM_001098723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020369.mm.1 | NM_001098723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020369.mm.1 | TC0X00003250.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437495_at | - | - |
| Mouse Expression Set 430 | 1437495_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 844323 | - | - |

Yy2
Genomic Map