Assay Details
| Gene Symbol: Klra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252577.1 | NP_001239506.1 | 2 | 203 | 79 | |
| NM_010649.3 | NP_034779.1 | 2 | 215 | 79 | ||
| GenBank mRNA | AK080171.1 | - | 2 | 204 | 79 | |
| AK145141.1 | - | 1 | 74 | 79 | ||
| AY860973.1 | - | 58 | 79 | |||
| L78247.1 | - | 1 | 58 | 79 | ||
| U10090.1 | - | 1 | 125 | 79 | ||
| Gene Symbol: Klra18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053153.2 | NP_444383.1 | 2 | 202 | 79 | |
| GenBank mRNA | AF288377.1 | - | 2 | 202 | 79 | |
| BC111866.1 | - | 2 | 152 | 40052250 | 79 | |
| BC141242.1 | - | 1 | 150 | 9055854 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998006 | NM_001039118.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998006 | AY860973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004536 | U10090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004536 | NM_010649.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004536 | AK080171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004536 | AK145141.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998006 | NM_001039118.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998006 | AY860973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004536 | U10090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004536 | NM_010649.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004536 | AK080171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004536 | AK145141.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24846423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24846444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24846431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24846450 | - | - |
| Whole Mouse Genome G4122A | A_51_P306387 | NM_010649.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P306387 | AK080171.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24846396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05292003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05658070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05031954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05031947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05031957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05082726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05305597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24846415 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548487 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548463 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548465 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548465 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548463 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548464 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426127_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608638 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548455 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548455 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548455 | - | - |
| Mouse Genome 430 2.0 Array | 1426127_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1426127_PM_x_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa13040_f_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600025469.mm.1 | NM_001252577.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025469.mm.1 | NM_001252577.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025469.mm.1 | NM_010649.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025469.mm.1 | NM_010649.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025469.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025448.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025464.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025468.mm.1 | NM_001252577.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025468.mm.1 | NM_001252577.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025468.mm.1 | NM_010649.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025468.mm.1 | NM_010649.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025468.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025462.mm.1 | NM_001252577.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025462.mm.1 | NM_001252577.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025462.mm.1 | NM_010649.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025462.mm.1 | NM_010649.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025462.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025463.mm.1 | NM_001252577.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025463.mm.1 | NM_001252577.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025463.mm.1 | NM_010649.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025463.mm.1 | NM_010649.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025463.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025447.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025449.mm.1 | TC0600003607.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426127_PM_x_at | - | - |
| Mouse Expression Set 430 | 1426127_x_at | - | - |
| Murine Genome U74v2 Set | 100387_f_at | - | - |
| Murine Genome U74v2 Set | 100387_PM_f_at | - | - |
| Murine Genome U74v2 Set | 94776_f_at | - | - |
| Murine Genome U74v2 Set | 100387_PM_f_at | - | - |
| Murine Genome U74v2 Set | 95908_PM_at | - | - |
| Murine Genome U74v2 Set | 100387_f_at | - | - |
| Murine Genome U74v2 Set | 100387_f_at | - | - |
| Murine Genome U74v2 Set | 95908_at | - | - |
| Murine Genome U74v2 Set | 100387_PM_f_at | - | - |
| Murine Genome U74v2 Set | 94776_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426127_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426127_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426127_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426127_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 806675 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1780736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104670687 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101400091 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1850253 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1340176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1780736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340176 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001780736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242590 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252295 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242590 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252295 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251366 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249056 | - | - |
| mouse-6 v1.1 (Illumina) | scl093970.1_92-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_21361219-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0016635.1_262-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_6754461-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0093970.1_84-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104670687 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101400091 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1780736 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850253 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340176 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0093970.1_84-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0016635.1_262-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl093970.1_92-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340176 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104670687 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101400091 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1780736 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850253 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3031402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2982019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1252295 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3104704 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251366 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2982019 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242590 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3104704 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252295 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3031402 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl093970.1_92-S | - | - |

Klra4
Genomic Map