Assay Details
| Gene Symbol: Vmn2r106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104568.1 | NP_001098038.1 | 3 - 4 | 1311 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988253 | NM_001104568.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901672 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230735 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 199064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835761 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841468 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40267 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988253 | NM_001104568.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31081992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31081998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31081996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31081993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31082073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13359131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13359129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13563172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31082119 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1700013609.mm.1 | NM_001104568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013609.mm.1 | NM_001104568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013609.mm.1 | TC1700001536.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013611.mm.1 | NM_001104568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013611.mm.1 | NM_001104568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013611.mm.1 | TC1700001536.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013614.mm.1 | NM_001104568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013614.mm.1 | NM_001104568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013614.mm.1 | TC1700001536.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013610.mm.1 | NM_001104568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013610.mm.1 | NM_001104568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013610.mm.1 | TC1700001536.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013612.mm.1 | NM_001104568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013612.mm.1 | NM_001104568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013612.mm.1 | TC1700001536.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013613.mm.1 | NM_001104568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013613.mm.1 | NM_001104568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013613.mm.1 | TC1700001536.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 520887 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215517 | - | - |

Vmn2r106
Genomic Map