Assay Details
| Gene Symbol: Cyp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136059.2 | NP_001129531.1 | 3 - 4 | 1066 | 131 | |
| NM_009992.4 | NP_034122.1 | 3 - 4 | 1145 | 131 | ||
| GenBank mRNA | AK005000.1 | - | 3 - 4 | 1146 | 131 | |
| BC125440.1 | - | 3 - 4 | 1065 | 40129955 | 131 | |
| BC125444.1 | - | 3 - 4 | 1065 | 40129959 | 131 | |
| K02588.1 | - | 3 - 4 | 1065 | 131 | ||
| Y00071.1 | - | 3 - 4 | 1066 | 131 | ||
More Information
Set Membership: |
|

Cyp1a1
Genomic Map