Assay Details
| Gene Symbol: Klra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010649.3 | NP_034779.1 | 2 | 244 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_010649.3 transcripts.
Set Membership: |
|

Klra4
Genomic Map