Assay Details
| Gene Symbol: Hbs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042593.1 | NP_001036058.1 | 2 - 3 | 265 | 94 | |
| NM_001145209.1 | NP_001138681.1 | 2 - 3 | 265 | 94 | ||
| NM_019702.2 | NP_062676.2 | 2 - 3 | 265 | 94 | ||
| GenBank mRNA | AK006626.1 | - | 2 - 3 | 238 | 94 | |
| AK012856.1 | - | 2 - 3 | 215 | 94 | ||
| AK146678.1 | - | 2 - 3 | 231 | 94 | ||
| AK166140.1 | - | 2 - 3 | 228 | 94 | ||
| AK168671.1 | - | 2 - 3 | 228 | 94 | ||
| AK169281.1 | - | 2 - 3 | 228 | 94 | ||
| AK173091.1 | - | 2 - 3 | 211 | 94 | ||
| BC010251.1 | - | 2 - 3 | 190 | 3964560 | 94 | |
| BC085235.1 | - | 2 - 3 | 207 | 5328213 | 94 | |
| BU510509.1 | - | 2 - 3 | 217 | 94 | ||
| CJ082265.1 | - | 2 - 3 | 265 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hbs1l
Genomic Map