Assay Details
| Gene Symbol: Has1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008215.2 | NP_032241.1 | 3 - 4 | 985 | 65 | |
| GenBank mRNA | AK138203.1 | - | 2 - 3 | 271 | 65 | |
| BC125424.1 | - | 3 - 4 | 964 | 40129939 | 65 | |
| BC125426.1 | - | 3 - 4 | 964 | 40129941 | 65 | |
| D82964.1 | - | 3 - 4 | 991 | 65 | ||
More Information
Set Membership: |
|

Has1
Genomic Map