Assay Details
| Gene Symbol: Srsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011358.2 | NP_035488.1 | 2 - 3 | 838 | 74 | |
| GenBank mRNA | AF077858.1 | - | 2 - 3 | 826 | 74 | |
| AF250132.1 | - | 1 - 2 | 111 | 74 | ||
| AK086103.1 | - | 2 - 3 | 836 | 74 | ||
| AK088042.1 | - | 2 - 3 | 836 | 74 | ||
| AK161873.1 | - | 2 - 3 | 834 | 74 | ||
| AK165670.1 | - | 2 - 3 | 840 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srsf2
Genomic Map