Assay Details
| Gene Symbol: 1700025N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040523.1 | 1 - 2 | 80 | 99 | ||
| GenBank mRNA | AK006361.1 | - | 1 - 2 | 80 | 99 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY706428.1 | - | Chr. - - [-] on Build | 99 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 38793 |
Gm38793 | -79.2kb | Sense | Chr. 6 - 38958260 - 38986574 on NCBI Build GRCm38 | |
| Downstream |
poly (ADP-ribose) polymerase family, member 12 |
Parp12 |
9930021O16, AA409132, AA536654, ARTD12, PARP-12, Zc3hdc1 |
18.8kb | Sense | Chr. 6 - 39086412 - 39118349 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700025N23Rik
Genomic Map