Assay Details
| Gene Symbol: 1700051K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006761.1 | - | 1 | 447 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 17089 | Gm17089 | Sense | Chr. 6 - 134233660 - 134248799 on NCBI Build GRCm38 | ||
| Downstream | BCL2-like 14 (apoptosis facilitator) | Bcl2l14 |
4930452K23Rik, 4933405K19Rik, 9030625M01Rik, AI429211, Bclg |
150.0kb | Anti-sense | Chr. 6 - 134396329 - 134438741 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700051K13Rik
Genomic Map