Assay Details
| Gene Symbol: 2300004M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009029.1 | - | 1 | 1148 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear receptor coactivator 2 | Ncoa2 |
D1Ertd433e, GRIP-1, Grip1, KAT13C, NCoA-2, SRC-2, TIF2, bHLHe75 |
Sense | Chr. 1 - 13139105 - 13374691 on NCBI Build GRCm38 | |
| Downstream | translocating chain-associating membrane protein 1 | Tram1 |
1810049E02Rik, TRAMP |
418.1kb | Sense | Chr. 1 - 13564693 - 13589910 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2300004M11Rik
Genomic Map