Assay Details
| Gene Symbol: Mrps18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034502.1 | - | 253 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | alpha tubulin acetyltransferase 1 | Atat1 |
0610011P08Rik, 2610008K08Rik, 2610110G12Rik, 3110080J08Rik, Mec17, TAT |
Anti-sense | Chr. 17 - 35897595 - 35910093 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Mrps18b
Genomic Map