Assay Details
| Gene Symbol: Mrps18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006558.1 | - | 359 | 63 | ||
| AK034502.1 | - | 931 | 63 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | alpha tubulin acetyltransferase 1 | Atat1 |
0610011P08Rik, 2610008K08Rik, 2610110G12Rik, 3110080J08Rik, Mec17, TAT |
Anti-sense | Chr. 17 - 35897595 - 35910093 on NCBI Build GRCm38 | |
| Downstream |
protein phosphatase 1, regulatory subunit 10 |
Ppp1r10 |
2610025H06Rik, Cat53, D17Ertd808e, Fb19, Pnuts |
6.0kb | Sense | Chr. 17 - 35916512 - 35952996 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrps18b
Genomic Map