Assay Details
| Gene Symbol: 6820402I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018358.1 | - | 1 | 2056 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin-conjugating enzyme E2E 1 | Ube2e1 |
UbcM3, Ubce5, ubcM2 |
-368.0kb | Sense | Chr. 14 - 18282726 - 18331844 on NCBI Build GRCm38 |
| Downstream | predicted gene 2237 | Gm2237 | ENSMUSG00000079354 | 861.8kb | Sense | Chr. 14 - 19563750 - 19584066 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6820402I19Rik
Genomic Map