Assay Details
| Gene Symbol: 8430437O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018474.1 | - | 1 | 154 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | peroxisomal biogenesis factor 19 | Pex19 | Pxf | -47.9kb | Sense | Chr. 1 - 172126744 - 172136497 on NCBI Build GRCm38 |
| Downstream | phosphoprotein enriched in astrocytes 15A | Pea15a |
Mat1, PEA-15, Pea15, Pkcs15 |
11.0kb | Anti-sense | Chr. 1 - 172196728 - 172206781 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

8430437O03Rik
Genomic Map