Assay Details
Gene Symbol: 2610028E06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015560.1 | 3 - 4 | 461 | 66 | ||
GenBank mRNA | AK031953.1 | - | 3 - 4 | 461 | 66 | |
AK082619.1 | - | 3 - 4 | 386 | 66 | ||
AK083757.1 | - | 4 - 5 | 874 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
glutamate receptor, ionotropic, kainate 3 |
Grik3 |
9630027E11, GluK3, GluR7-3, Glur-7, Glur7 |
-179.0kb | Anti-sense | Chr. 4 - 125490709 - 125714173 on NCBI Build GRCm38 |
Downstream | colony stimulating factor 3 receptor (granulocyte) | Csf3r |
Cd114, Csfgr, G-CSF-R, G-CSFR |
129.5kb | Anti-sense | Chr. 4 - 126023441 - 126044975 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
