Assay Details
| Gene Symbol: Gm7715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131765.1 | - | 348 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | choline acetyltransferase | Chat | B230380D24Rik | -47.4kb | Anti-sense | Chr. 14 - 32408203 - 32465909 on NCBI Build GRCm38 |
| Downstream | paired related homeobox protein-like 1 | Prrxl1 |
Drg11, Drgx |
82.3kb | Sense | Chr. 14 - 32595838 - 32649320 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm7715
Genomic Map