Assay Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009919.1 | - | 1 | 444 | 110 | |
| AK033430.1 | - | 1 | 2584 | 110 | ||
| AK081236.1 | - | 1 | 2134 | 110 | ||
| AK150620.1 | - | 1 | 954 | 110 | ||
| AK153354.1 | - | 1 | 954 | 110 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hippocampus abundant gene transcript 1 | Hiat1 | -7.6kb | Sense | Chr. 3 - 116631164 - 116662692 on NCBI Build GRCm38 | |
| Downstream |
amylo-1,6-glucosidase, 4-alpha-glucanotransferase |
Agl |
1110061O17Rik, 9430004C13Rik, 9630046L06Rik, AI850929, C77197 |
68.2kb | Sense | Chr. 3 - 116739999 - 116808166 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc35a3
Genomic Map