| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1453745_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1453745_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1453745_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0500001464.mm.1 |
NR_045040.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001464.mm.1 |
NR_045040.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001464.mm.1 |
NR_045042.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001464.mm.1 |
NR_045042.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001464.mm.1 |
TC0500000199.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001460.mm.1 |
NR_045040.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001460.mm.1 |
NR_045040.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001460.mm.1 |
NR_045042.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001460.mm.1 |
NR_045042.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001460.mm.1 |
TC0500000199.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001457.mm.1 |
NR_045040.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001457.mm.1 |
NR_045040.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001457.mm.1 |
NR_045042.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001457.mm.1 |
NR_045042.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001457.mm.1 |
TC0500000199.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001459.mm.1 |
NR_045040.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001459.mm.1 |
NR_045040.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001459.mm.1 |
NR_045042.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001459.mm.1 |
NR_045042.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001459.mm.1 |
TC0500000199.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001463.mm.1 |
NR_045040.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001463.mm.1 |
NR_045040.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001463.mm.1 |
NR_045042.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001463.mm.1 |
NR_045042.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001463.mm.1 |
TC0500000199.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1453745_at |
- |
- |
| Mouse Expression Set 430 |
1453745_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453745_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453745_PM_at |
- |
- |