Assay Details
| Gene Symbol: Ccny | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014507.1 | - | 1 | 1246 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
gap junction protein, delta 4 |
Gjd4 |
9430022F06Rik, Cx39 |
-22.5kb | Sense | Chr. 18 - 9137290 - 9289808 on NCBI Build GRCm38 |
| Downstream | centrin 1 | Cetn1 | caltractin | 303.5kb | Sense | Chr. 18 - 9618419 - 9619469 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccny
Genomic Map