Assay Details
| Gene Symbol: 4833428M15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014787.1 | - | 1 | 1448 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dachshund 1 (Drosophila) | Dach1 |
Dac, Dach |
Sense | Chr. 14 - 97786846 - 98169765 on NCBI Build GRCm38 | |
| Downstream | mitotic spindle organizing protein 1 | Mzt1 |
2410129H14Rik, AI120158, Mozart1 |
947.8kb | Sense | Chr. 14 - 99034544 - 99046463 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4833428M15Rik
Genomic Map