Assay Details
| Gene Symbol: 1110015O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045272.1 | 4 | 1036 | 106 | ||
| GenBank mRNA | AK003731.2 | - | 1 | 552 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
hepatocyte nuclear factor 4, gamma |
Hnf4g | NR2A2 | -1.2mb | Sense | Chr. 3 - 3508030 - 3659800 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 17308 |
Gm17308 | 275.4kb | Sense | Chr. 3 - 5090276 - 5093402 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1110015O18Rik
Genomic Map