Assay Details
| Gene Symbol: 2900060B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027901.1 | 1 | 67 | 72 | ||
| GenBank mRNA | AK019334.1 | - | 1 | 68 | 72 | |
| AK019335.1 | - | 1 | 68 | 72 | ||
| AK019348.1 | - | 1 | 68 | 72 | ||
| AK050598.1 | - | 1 | 67 | 72 | ||
| AK083371.1 | - | 1 | 67 | 72 | ||
| AK169832.1 | - | 1 | 68 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 28867 | Gm28867 | -96.2kb | Anti-sense | Chr. 1 - 118359290 - 118362918 on NCBI Build GRCm38 | |
| Downstream | transcription factor CP2-like 1 | Tfcp2l1 |
1810030F05Rik, 4932442M07Rik, AA575098, Cp2l1, Crtr-1, D930018N21Rik, LBP-9, Tcfcp2l1 |
168.7kb | Anti-sense | Chr. 1 - 118627923 - 118685168 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK169832.1 transcripts.
This assay is in the 5'UTR region of the AK169832.1 transcripts.
Set Membership: |
|

2900060B14Rik
Genomic Map