Assay Details
| Gene Symbol: 9330154F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020373.1 | - | 1 | 149 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bone morphogenic protein/retinoic acid inducible neural specific 1 | Brinp1 |
BRINP, Dbc1, Dbccr1, Fam5a |
Sense | Chr. 4 - 68761372 - 68954397 on NCBI Build GRCm38 | |
| Downstream | stathmin 1 pseudogene | Gm11223 | OTTMUSG00000000267 | 1.1mb | Sense | Chr. 4 - 69925201 - 69926170 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9330154F10Rik
Genomic Map