Assay Details
| Gene Symbol: 2610024D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011527.1 | - | 1 | 653 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 40824 |
Gm40824 | Sense | Chr. 11 - 21050355 - 21110192 on NCBI Build GRCm38 | ||
| Downstream | vacuolar protein sorting 54 (yeast) | Vps54 |
Hcc8, Vps54l, mSLP8, wr |
138.4kb | Sense | Chr. 11 - 21239032 - 21321133 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610024D14Rik
Genomic Map