Assay Details
| Gene Symbol: C530020B09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017534.1 | - | 1 | 1156 | 80 | |
| AK049667.1 | - | 1 | 1208 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
amylase 1, salivary |
Amy1 |
Amy-1, Amy1a, C030014B17Rik |
-52.2kb | Anti-sense | Chr. 3 - 113555952 - 113577750 on NCBI Build GRCm38 |
| Downstream |
collagen, type XI, alpha 1 |
Col11a1 |
C530001D20Rik, cho |
399.2kb | Sense | Chr. 3 - 114030479 - 114220327 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C530020B09Rik
Genomic Map