Assay Details
Gene Symbol: Gm33466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139694.1 | - | 3 | 1345 | 114 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | prostaglandin E receptor 3 (subtype EP3) | Ptger3 |
EP3, Pgerep3, Ptgerep3 |
-75.6kb | Anti-sense | Chr. 3 - 157566816 - 157644758 on NCBI Build GRCm38 |
Downstream | cystathionase (cystathionine gamma-lyase) | Cth |
0610010I13Rik, AI314617, BC019483, CSE, Cys3 |
173.2kb | Sense | Chr. 3 - 157894248 - 157925081 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
