Assay Details
| Gene Symbol: Lpar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172989.1 | NP_766577.1 | 1 - 2 | 160 | 82 | |
| GenBank mRNA | AK030330.1 | - | 1 - 2 | 123 | 83 | |
| BC025425.1 | - | 1 - 2 | 70 | 5009055 | 82 | |
| BY716867.1 | - | 1 - 2 | 119 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030330.1, BC025425.1, NM_172989.1 transcripts.
Set Membership: |
|

Lpar1
Genomic Map