| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300001237.mm.1 |
NM_001162928.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001237.mm.1 |
NM_001162928.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001237.mm.1 |
NM_001162929.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001237.mm.1 |
NM_001162929.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001237.mm.1 |
TC1300000187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300001236.mm.1 |
NM_001162928.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001236.mm.1 |
NM_001162928.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001236.mm.1 |
NM_001162929.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001236.mm.1 |
NM_001162929.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001236.mm.1 |
TC1300000187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300001242.mm.1 |
NM_001162929.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001242.mm.1 |
NM_001162929.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300001242.mm.1 |
TC1300000187.mm.1 |
MTA TC |