Assay Details
| Gene Symbol: Olfr420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146305.2 | NP_666417.2 | 1 | 651 | 71 | |
| GenBank mRNA | BC145885.1 | - | 1 | 651 | 40131188 | 71 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372372 | NM_146305.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372372 | BC145885.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372372 | NM_146305.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372372 | BC145885.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302724 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142008 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372372 | NM_146305.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372372 | BC145885.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372372 | NM_146305.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372372 | BC145885.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372372 | NM_146305.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372372 | BC145885.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372372 | NM_146305.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372372 | BC145885.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20841218 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142008 | - | - |
| Whole Mouse Genome G4122A | A_51_P372372 | NM_146305.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372372 | BC145885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372372 | NM_146305.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372372 | BC145885.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130104 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142008 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00739327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00425166 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00500049 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00500048 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00500044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00134137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00910591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00771205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00500047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00500044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00500049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00503039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00771204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841187 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351904 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351903 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100013622.mm.1 | NM_146305.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013622.mm.1 | NM_146305.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013622.mm.1 | TC0100001643.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 364772 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2640402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640402 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241294 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241294 | - | - |
| mouse-6 v1.1 (Illumina) | scl14316.1.1_77-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640402 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl14316.1.1_77-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1241294 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241294 | - | - |

Olfr420
Genomic Map