Assay Details
| Gene Symbol: Olfr902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146802.2 | NP_667013.2 | 1 | 44 | 100 | |
| GenBank mRNA | BC145901.1 | - | 1 | 44 | 40131204 | 100 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC145901.1, NM_146802.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957802 | BC145901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957802 | NM_146802.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371893 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154478 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957802 | BC145901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957802 | NM_146802.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P205772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434742 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154478 | - | - |
| Whole Mouse Genome G4122A | A_52_P205772 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109430 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154478 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479835 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479821 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08101419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26434752 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584410 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584409 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900002799.mm.1 | NM_146802.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002799.mm.1 | NM_146802.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002799.mm.1 | TC0900000400.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1240195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2992434 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2992434 | - | - |

Olfr902
Genomic Map