| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400016657.mm.1 |
NM_001164329.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016657.mm.1 |
NM_001164329.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016657.mm.1 |
TC1400002160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400016659.mm.1 |
NM_001164329.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016659.mm.1 |
NM_001164329.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016659.mm.1 |
TC1400002160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400016658.mm.1 |
NM_001164329.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016658.mm.1 |
NM_001164329.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016658.mm.1 |
TC1400002160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400016662.mm.1 |
NM_001164329.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016662.mm.1 |
NM_001164329.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016662.mm.1 |
TC1400002160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400016660.mm.1 |
NM_001164329.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016660.mm.1 |
NM_001164329.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016660.mm.1 |
TC1400002160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400016661.mm.1 |
NM_001164329.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016661.mm.1 |
NM_001164329.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400016661.mm.1 |
TC1400002160.mm.1 |
MTA TC |