Assay Details
| Gene Symbol: Cdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166658.1 | NP_001160130.1 | 2151 | 82 | ||
More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC0X00002320.mm
This assay targets the same location as RTA transcript cluster(s) TC0X00002534.rn
This assay targets the same location as RTA transcript cluster(s) TC0X00002534.rn
Set Membership: |
|

Cdr1
Genomic Map