Assay Details
| Gene Symbol: Olfr320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207230.1 | NP_997113.1 | 1 | 49 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356842 | NM_207230.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356842 | NM_207230.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356842 | NM_207230.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356842 | NM_207230.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356842 | NM_207230.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356842 | NM_207230.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27832266 | - | - |
| Whole Mouse Genome G4122A | A_51_P356842 | NM_207230.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356842 | NM_207230.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092815 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09254124 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092813 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092814 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09092819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27832322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09092811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09355608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09104234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09045327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09104235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09045325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09254117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27832277 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10386208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376405 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376409 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376404 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10386207 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376408 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100005589.mm.1 | NM_207230.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005589.mm.1 | NM_207230.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005589.mm.1 | TC1100000697.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 897398 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 106220433 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 70364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070364 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2535194 | - | - |
| mouse-6 v1.1 (Illumina) | GI_46402210-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0216783.1_229-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70364 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106220433 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0216783.1_229-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70364 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106220433 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2949449 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2949449 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236773 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2535194 | - | - |

Olfr320
Genomic Map