Assay Details
| Gene Symbol: Olfr1030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146588.2 | NP_666799.1 | 2 | 855 | 92 | |
| GenBank mRNA | BC128001.1 | - | 1 | 826 | 40135864 | 92 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr1030
Genomic Map