| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10577439 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10577427 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10577428 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10577423 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10577438 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10577431 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10577440 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10577436 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10577422 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10577430 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1429378_PM_x_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10577429 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10577426 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10577437 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10577434 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10577421 |
- |
- |
| Mouse Genome 430 2.0 Array |
1429378_PM_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1429378_x_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
382000_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
382000_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0800014615.mm.1 |
NM_001007582.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014615.mm.1 |
NM_001007582.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014615.mm.1 |
TC0800001851.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800014616.mm.1 |
NM_001007582.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014616.mm.1 |
NM_001007582.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014616.mm.1 |
TC0800001851.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1429378_x_at |
- |
- |
| Mouse Expression Set 430 |
1429378_PM_x_at |
- |
- |
| Murine Genome U74v2 Set |
167227_r_at |
- |
- |
| Murine Genome U74v2 Set |
167227_PM_r_at |
- |
- |
| Murine Genome U74v2 Set |
165941_at |
- |
- |
| Murine Genome U74v2 Set |
165941_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1429378_PM_x_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1429378_x_at |
- |
- |