Assay Details
| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019436.3 | NP_062309.2 | 4 - 5 | 437 | 100 | |
| GenBank mRNA | AJ236881.1 | - | 4 - 5 | 413 | 100 | |
| AK079882.1 | - | 4 - 5 | 439 | 100 | ||
| BC064055.1 | - | 4 - 5 | 450 | 576098 | 100 | |
More Information
Set Membership: |
|

Sit1
Genomic Map