Assay Details
| Gene Symbol: Dyrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014390.2 | NP_001014412.1 | 1 - 2 | 182 | 79 | |
| GenBank mRNA | AK149371.1 | - | 1 - 2 | 79 | ||
| BC085145.1 | - | 1 - 2 | 182 | 6808145 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085145.1, NM_001014390.2 transcripts.
Set Membership: |
|

Dyrk2
Genomic Map