Assay Details
| Gene Symbol: Pcdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042726.3 | NP_001036191.1 | 1 - 2 | 2524 | 105 | |
| GenBank mRNA | AK048638.1 | - | 1 - 2 | 2525 | 105 | |
| AK140407.1 | - | 1 - 2 | 2412 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcdh8
Genomic Map