Assay Details
| Gene Symbol: Pcdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042726.3 | NP_001036191.1 | 2 - 3 | 2738 | 84 | |
| NM_021543.4 | NP_067518.2 | 2 - 3 | 3029 | 84 | ||
| GenBank mRNA | AF231125.1 | - | 2 - 3 | 2953 | 84 | |
| AK048638.1 | - | 2 - 3 | 2739 | 84 | ||
| AK140407.1 | - | 2 - 3 | 2626 | 84 | ||
| BC053008.1 | - | 2 - 3 | 3017 | 5698009 | 84 | |
More Information
Set Membership: |
|

Pcdh8
Genomic Map